Product References
Fisetin alleviates sepsis-induced multiple organ dysfunction in mice via inhibiting p38 MAPK/MK2 signaling.
Acta pharmacologica Sinica
Zhang HF,Zhang HB,Wu XP,Guo YL,Cheng WD,Qian F
RM3001 was used in Flow cytometry/Cell sorting to demonstrate that fisetin is a promising agent for protecting against sepsis-induced inflammatory response and organ injury via inhibiting macrophage activation.
Thu Oct 01 00:00:00 EDT 2020
Autofluorescence multiphoton microscopy for visualization of tissue morphology and cellular dynamics in murine and human airways.
Laboratory investigation; a journal of technical methods and pathology
Kretschmer S,Pieper M,Hüttmann G,Bölke T,Wollenberg B,Marsh LM,Garn H,König P
RM3001 was used in immunohistochemistry to propose that autofluorescence-based imaging finds previously unknown immune cell interactions in the airway
Mon Aug 01 00:00:00 EDT 2016
I/St mice hypersusceptible to Mycobacterium tuberculosis are resistant to M. avium.
Infection and immunity
Kondratieva EV,Evstifeev VV,Kondratieva TK,Petrovskaya SN,Pichugin AV,Rubakova EI,Averbakh MM,Apt AS
RM3001 was used in Flow cytometry/Cell sorting to study the infection caused by Mycobacterium avium virulent strain 724 in a panel of inbred mouse strains and found that I/St mice are resistant to M. avium.
Mon Oct 01 00:00:00 EDT 2007
Hematopoiesis and stem cell renewal in long-term bone marrow cultures containing catalase.
Blood
Gupta R,Karpatkin S,Basch RS
RM3001 was used in flow cytometry to investigate how peroxide affects hematopoiesis
Wed Mar 01 00:00:00 EST 2006
Hydrolytic and nonenzymatic functions of acetylcholinesterase comodulate hemopoietic stress responses.
Journal of immunology (Baltimore, Md. : 1950)
Grisaru D,Pick M,Perry C,Sklan EH,Almog R,Goldberg I,Naparstek E,Lessing JB,Soreq H,Deutsch V
RM3001 was used in flow cytometry to demonstrate that AChE-R and ARP facilitate granulocytosis
Sun Jan 01 00:00:00 EST 2006
Mycobacterial dissemination and cellular responses after 1-lobe restricted tuberculosis infection of genetically susceptible and resistant mice.
The Journal of infectious diseases
Mischenko VV,Kapina MA,Eruslanov EB,Kondratieva EV,Lyadova IV,Young DB,Apt AS
RM3001 was used in flow cytometry to study mycobacterial dissemination and immune-cell trafficking in tuberculosis
Wed Dec 15 00:00:00 EST 2004
Schistosoma mansoni worms induce anergy of T cells via selective up-regulation of programmed death ligand 1 on macrophages.
Journal of immunology (Baltimore, Md. : 1950)
Smith P,Walsh CM,Mangan NE,Fallon RE,Sayers JR,McKenzie AN,Fallon PG
RM3001 was used in flow cytometry to elucidate how Schistosoma mansoni suppresses T cell activation
Thu Jul 15 00:00:00 EDT 2004
Reduced apoptosis and ameliorated listeriosis in TRAIL-null mice.
Journal of immunology (Baltimore, Md. : 1950)
Zheng SJ,Jiang J,Shen H,Chen YH
RM3001 was used in flow cytometry to identify a role for TRAIL in apoptosis during listeriosis
Mon Nov 01 00:00:00 EST 2004
Myeloid cell expansion elicited by the progression of spontaneous mammary carcinomas in c-erbB-2 transgenic BALB/c mice suppresses immune reactivity.
Blood
Melani C,Chiodoni C,Forni G,Colombo MP
RM3001 was used in flow cytometry to use the BALB-neuT model to study carcinogenesis.
Mon Sep 15 00:00:00 EDT 2003
Extensive proliferation of T cell lineage-restricted progenitors in the thymus: an essential process for clonal expression of diverse T cell receptor beta chains.
European journal of immunology
Kawamoto H,Ohmura K,Fujimoto S,Lu M,Ikawa T,Katsura Y
RM3001 was used in flow cytometry to assess the proliferative potential of thymic progenitors that are T cell lineage restricted.
Sat Mar 01 00:00:00 EST 2003
Passive transfer of flt-3L-derived dendritic cells delays diabetes development in NOD mice and associates with early production of interleukin (IL)-4 and IL-10 in the spleen of recipient mice.
Clinical and experimental immunology
Morin J,Faideau B,Gagnerault MC,Lepault F,Boitard C,Boudaly S
RM3001 was used in flow cytometry to assess the contribution of DC function in non-obese diabetic mice.
Mon Dec 01 00:00:00 EST 2003
Downmodulation of the inflammatory response to bacterial infection by gammadelta T cells cytotoxic for activated macrophages.
The Journal of experimental medicine
Egan PJ,Carding SR
RM3001 was used in flow cytometry to propose that gammadelta T cells eliminating activated macrophages after infection with an intracellular pathogen.
Mon Jun 19 00:00:00 EDT 2000
Ly-6C is a monocyte/macrophage and endothelial cell differentiation antigen regulated by interferon-gamma.
European journal of immunology
Jutila MA,Kroese FG,Jutila KL,Stall AM,Fiering S,Herzenberg LA,Berg EL,Butcher EC
RM3001 was used in flow cytometry to use a new Ly-6C-specific antibody to study monocytes, macrophages, and endothelial cells
Tue Nov 01 00:00:00 EST 1988
Leukocyte-endothelial cell recognition: evidence of a common molecular mechanism shared by neutrophils, lymphocytes, and other leukocytes.
Journal of immunology (Baltimore, Md. : 1950)
Lewinsohn DM,Bargatze RF,Butcher EC
RM3001 was used in flow cytometry to show the involvement of the MEL-14 antigen in neutrophil-endothelial cell interactions
Mon Jun 15 00:00:00 EDT 1987