Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 13
Retinoid-related orphan receptor beta (ROR beta), a NR1 thyroid hormone receptor, affects the development and functioning of the central nervous system, including sensory input integration and circadian timing. Disruption of ROR beta in mice results in juvenile ataxia, retinal degeneration, circadian activity abnormalities, and delayed onset of male fertility. ROR beta binds to the monomeric response elements containing the sequence AnnTAGGTCA as a monomer. An alternatively spliced isoform, ROR beta2, has been isolated from rats, and the expression of ROR beta2 is confined to the pineal gland and retina. ROR beta2 shares common DNA- and putative ligand-binding domains with wild-type ROR beta but is characterized by a different amino-terminal domain. ROR beta expression has been documented in various regions of the mouse brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: nuclear receptor RZR-beta; nuclear receptor subfamily 1 group F member 2; RAR-related orphan receptor B; RAR-related orphan receptor beta; retinoic acid-binding receptor beta; retinoid-related orphan receptor beta; retinoid-related orphan receptor-beta
Gene Aliases: bA133M9.1; NR1F2; ROR-BETA; RZR-BETA; RZRB
Entrez Gene ID: (Human) 6096
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support