Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
CD160
Entrez Gene ID
11126
Gene Name:
CD160 molecule
Gene Aliases
BY55, NK1, NK28
Gene Chromosome Location
Chr.1: on Build GRCh38
UniGene
Hs.488237
Species:
Human
Primer Chromosome Location
Chr.1: 145731198-145730671 on Build GRCh38
View in UCSC browser
Amplicon Length
528
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00399974_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
CD160
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_007053.3
NP_008984.1
5377514
c.242G>A
p.R81K
5553721
c.211G>C
p.E71Q
5377515
c.164G>A
p.G55E
4639731
c.114G>A
p.T38T
5917045
c.400+8C>T
p.?
5911244
c.196G>A
p.G66R
895248
c.115C>T
p.R39*
6121041
c.241A>G
p.R81G
5930440
c.400+1G>A
p.?
895245
c.371A>G
p.Q124R
5887175
c.364C>A
p.R122S
895247
c.151G>T
p.E51*
895246
c.365G>A
p.R122H
6873450
c.218G>A
p.S73N
1200227
c.368T>G
p.L123R
1626388
c.251G>A
p.G84E
6880512
c.310C>T
p.P104S
1688471
c.268G>A
p.E90K
6623407
c.311C>T
p.P104L
6623409
c.112A>G
p.T38A
6623408
c.314T>C
p.L105S
3735930
c.277T>A
p.S93T
3473163
c.110G>A
p.G37E
4004780
c.259G>A
p.G87S
2121232
c.364C>T
p.R122C
2121234
c.197G>T
p.G66V
279703
c.167T>G
p.F56C
NR_103845.1
-
-
-
XM_005272929.3
XP_005272986.1
-
-
-
XM_011509104.2
XP_011507406.1
-
-
-
GenBank
AF060981.1
AAC72302.1
-
-
-
AK128370.1
-
-
-
BC014465.1
AAH14465.1
-
-
-
CD638675.1
-
-
-
CR541867.1
CAG46665.1
-
-
-
CR541888.1
CAG46686.1
-
-
-
ENST00000369288.0
-
-
-
EU016100.1
-
-
-
EU016101.1
-
-
-
Phenotype
MIM: 604463
Literature Links:
CD160
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
cellular defense response
cell surface receptor signaling pathway
cell proliferation
regulation of immune response
defense response to Gram-negative bacterium
receptor activity
receptor binding
protein binding
MHC class I receptor activity
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9zxgn:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0