Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
GNRHR2
RBM8A
Entrez Gene ID
114814
9939
Gene Name:
gonadotropin releasing hormone receptor 2 (pseudogene)
RNA binding motif protein 8A
Gene Aliases
GnRH-II-R
BOV-1A, BOV-1B, BOV-1C, C1DELq21.1, DEL1q21.1, MDS014, RBM8, RBM8B, TAR, Y14, ZNRP, ZRNP1
Gene Chromosome Location
Chr.1: on Build GRCh38
Chr.1: on Build GRCh38
UniGene
Hs.356873
Hs.591455
Species:
Human
Primer Chromosome Location
Chr.1: 145922814-145922266 on Build GRCh38
View in UCSC browser
Amplicon Length
549
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00486328_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
GNRHR2
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NR_002328.4
-
-
-
NR_104033.1
-
-
-
GenBank
AY077708.1
-
-
-
AY081843.1
AAL89821.1
-
-
-
AY233133.1
AAP74658.1
-
-
-
ENST00000312753.0
-
-
-
Phenotype
MIM: 612875
RBM8A
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_005105.4
NP_005096.1
-
-
-
GenBank
AL049219.1
-
-
-
BC071577.1
-
-
-
Phenotype
MIM: 605313
Literature Links:
GNRHR2
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
cellular response to gonadotropin-releasing hormone
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
regulation of alternative mRNA splicing, via spliceosome
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
regulation of translation
mRNA 3'-end processing
gonadotropin-releasing hormone receptor activity
peptide binding
nucleotide binding
RNA binding
mRNA binding
protein binding
poly(A) RNA binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-55874ff697-s6fgm:80/10.239.167.51:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0