Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
WDR20 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001242414.1 | NP_001229343.1 | - | - | - |
NM_001242415.1 | NP_001229344.1 | - | - | - | |
NM_001242416.1 | NP_001229345.1 | - | - | - | |
NM_001242417.1 | NP_001229346.1 | 4243484 | c.848T>C | p.V283A | |
3493899 | c.1064C>T | p.P355L | |||
4243485 | c.891G>T | p.L297L | |||
5549491 | c.1025C>T | p.A342V | |||
4243483 | c.828G>T | p.R276R | |||
4406030 | c.1061A>G | p.D354G | |||
7271380 | c.866T>C | p.M289T | |||
7163920 | c.867G>C | p.M289I | |||
5913635 | c.992G>A | p.G331D | |||
4243490 | c.967G>C | p.V323L | |||
5740214 | c.1057G>T | p.G353C | |||
3706127 | c.1021G>A | p.V341I | |||
7132347 | c.859G>A | p.G287S | |||
4243487 | c.909G>A | p.P303P | |||
4243488 | c.924C>T | p.I308I | |||
4740887 | c.1040delC | p.T348fs*3 | |||
6035100 | c.828G>C | p.R276R | |||
6821616 | c.931G>A | p.G311S | |||
6501142 | c.846A>T | p.S282S | |||
4150723 | c.864G>A | p.T288T | |||
6458571 | c.997delG | p.H334fs*8 | |||
4147826 | c.936G>C | p.G312G | |||
NM_001242418.1 | NP_001229347.1 | - | - | - | |
NM_001320130.1 | NP_001307059.1 | - | - | - | |
NM_144574.3 | NP_653175.2 | - | - | - | |
NM_181291.2 | NP_851808.1 | - | - | - | |
NM_181308.2 | NP_851825.1 | - | - | - | |
NR_038354.1 | - | - | - | ||
XM_006720308.1 | XP_006720371.1 | - | - | - | |
XM_006720309.1 | XP_006720372.1 | - | - | - | |
XM_006720310.1 | XP_006720373.1 | - | - | - | |
XM_006720311.1 | XP_006720374.1 | - | - | - | |
XM_006720313.2 | XP_006720376.1 | - | - | - | |
XM_006720318.2 | XP_006720381.1 | - | - | - | |
XM_006720320.3 | XP_006720383.1 | - | - | - | |
XM_011537335.2 | XP_011535637.1 | - | - | - | |
XM_011537336.2 | XP_011535638.1 | - | - | - | |
XM_011537337.2 | XP_011535639.1 | - | - | - | |
XM_011537338.1 | XP_011535640.1 | - | - | - | |
XM_011537339.2 | XP_011535641.1 | - | - | - | |
XM_011537340.2 | XP_011535642.1 | - | - | - | |
XM_011537341.2 | XP_011535643.1 | - | - | - | |
XM_011537343.1 | XP_011535645.1 | - | - | - | |
XM_011537344.2 | XP_011535646.1 | - | - | - | |
XM_011537345.1 | XP_011535647.1 | - | - | - | |
XM_011537346.1 | XP_011535648.1 | - | - | - | |
XM_011537347.1 | XP_011535649.1 | - | - | - | |
XM_011537352.2 | XP_011535654.1 | - | - | - | |
XM_011537353.2 | XP_011535655.1 | - | - | - | |
XM_011537355.2 | XP_011535657.1 | - | - | - | |
XM_011537356.2 | XP_011535658.1 | 5549492 | c.932C>T | p.A311V | |
2024942 | c.735G>T | p.R245R | |||
2024943 | c.755T>C | p.V252A | |||
4147827 | c.843G>C | p.G281G | |||
5913636 | c.899G>A | p.G300D | |||
7271381 | c.773T>C | p.M258T | |||
5740215 | c.964G>T | p.G322C | |||
7163921 | c.774G>C | p.M258I | |||
248319 | c.850G>A | p.D284N | |||
2024950 | c.874G>C | p.V292L | |||
1300402 | c.766G>A | p.G256S | |||
2024947 | c.816G>A | p.P272P | |||
4406031 | c.968A>G | p.D323G | |||
2024948 | c.831C>T | p.I277I | |||
953520 | c.918G>T | p.W306C | |||
2024944 | c.771G>A | p.T257T | |||
6035101 | c.735G>C | p.R245R | |||
2024945 | c.798G>T | p.L266L | |||
1607344 | c.928G>A | p.V310I | |||
3493900 | c.971C>T | p.P324L | |||
6821617 | c.838G>A | p.G280S | |||
347019 | c.744C>G | p.N248K | |||
6501143 | c.753A>T | p.S251S | |||
4740888 | c.947delC | p.T317fs*3 | |||
6458572 | c.904delG | p.H303fs*8 | |||
XM_011537358.2 | XP_011535660.1 | - | - | - | |
XM_017021766.1 | XP_016877255.1 | - | - | - | |
XM_017021767.1 | XP_016877256.1 | - | - | - | |
XM_017021768.1 | XP_016877257.1 | - | - | - | |
XM_017021770.1 | XP_016877259.1 | - | - | - | |
XM_017021771.1 | XP_016877260.1 | - | - | - | |
XM_017021772.1 | XP_016877261.1 | - | - | - | |
XM_017021773.1 | XP_016877262.1 | - | - | - | |
XM_017021774.1 | XP_016877263.1 | - | - | - | |
XM_017021775.1 | XP_016877264.1 | - | - | - | |
XM_017021776.1 | XP_016877265.1 | - | - | - | |
XM_017021777.1 | XP_016877266.1 | - | - | - | |
XM_017021778.1 | XP_016877267.1 | - | - | - | |
XM_017021779.1 | XP_016877268.1 | - | - | - | |
XM_017021780.1 | XP_016877269.1 | - | - | - | |
XM_017021781.1 | XP_016877270.1 | - | - | - | |
XM_017021782.1 | XP_016877271.1 | - | - | - | |
XM_017021783.1 | XP_016877272.1 | - | - | - | |
XM_017021784.1 | XP_016877273.1 | - | - | - | |
XM_017021785.1 | XP_016877274.1 | - | - | - | |
GenBank | AB209148.1 | BAD92385.1 | - | - | - |
AF327354.1 | AAL56014.1 | - | - | - | |
AK056330.1 | - | - | - | ||
AK090580.1 | - | - | - | ||
AK090978.1 | - | - | - | ||
AK225852.1 | - | - | - | ||
AK297727.1 | - | - | - | ||
AL133558.1 | CAB63713.1 | - | - | - | |
AL832242.1 | - | - | - | ||
AL832500.1 | CAI46118.1 | - | - | - | |
BC028387.2 | AAH28387.1 | - | - | - | |
BC030654.2 | AAH30654.1 | - | - | - | |
BM707311.1 | - | - | - | ||
BX248274.1 | CAD62602.1 | - | - | - | |
DR423616.1 | - | - | - | ||
ENST00000335263.0 | - | - | - |
Literature Links: |
WDR20 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |