Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CDK8 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001260.2 | NP_001251.1 | - | - | - |
NM_001318368.1 | NP_001305297.1 | 4632056 | c.584T>C | p.V195A | |
6553855 | c.519C>T | p.D173D | |||
459306 | c.591C>T | p.F197F | |||
6980588 | c.646+1G>A | p.? | |||
1200578 | c.598C>T | p.R200* | |||
6944484 | c.569T>C | p.L190S | |||
1166001 | c.517G>A | p.D173N | |||
1677566 | c.594G>T | p.W198C | |||
6283865 | c.622G>T | p.A208S | |||
2069801 | c.518A>G | p.D173G | |||
6935844 | c.613C>T | p.L205F | |||
6969616 | c.515-2A>T | p.? | |||
6936256 | c.601G>T | p.A201S | |||
1366129 | c.532C>T | p.R178* | |||
946371 | c.557C>T | p.P186L | |||
6215503 | c.560T>C | p.L187S | |||
5376570 | c.574C>T | p.P192S | |||
1559411 | c.578T>G | p.V193G | |||
3936280 | c.533G>A | p.R178Q | |||
6922039 | c.584T>A | p.V195D | |||
5376569 | c.568T>G | p.L190V | |||
1247946 | c.613_615delCTT | p.L206delL | |||
3467991 | c.556C>T | p.P186S | |||
12811 | c.565G>A | p.D189N | |||
5761609 | c.595T>C | p.Y199H | |||
XM_005266216.4 | XP_005266273.1 | - | - | - | |
XM_011534864.1 | XP_011533166.1 | - | - | - | |
XM_011534865.2 | XP_011533167.1 | - | - | - | |
XM_017020317.1 | XP_016875806.1 | - | - | - | |
XM_017020318.1 | XP_016875807.1 | - | - | - | |
GenBank | AK296084.1 | - | - | - | |
AK296875.1 | - | - | - | ||
AY702976.1 | AAU29339.1 | - | - | - | |
BC069634.1 | AAH69634.1 | - | - | - | |
BC094784.1 | - | - | - | ||
BC104492.1 | AAI04493.1 | - | - | - | |
BC105950.1 | AAI05951.1 | - | - | - | |
BC107601.1 | AAI07602.1 | - | - | - | |
ENST00000381527.0 | - | - | - | ||
HG518164.1 | - | - | - | ||
X85753.1 | CAA59754.1 | - | - | - |
Literature Links: |
CDK8 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |