Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAPRIN2 | |||||
---|---|---|---|---|---|
Accession | Protein | COSMIC ID | Mutation | AA | |
RefSeq | NM_001002259.2 | NP_001002259.1 | - | - | - |
NM_001206856.2 | NP_001193785.1 | - | - | - | |
NM_001319842.1 | NP_001306771.1 | - | - | - | |
NM_001319843.1 | NP_001306772.1 | - | - | - | |
NM_001319844.1 | NP_001306773.1 | - | - | - | |
NM_001319845.1 | NP_001306774.1 | - | - | - | |
NM_001319846.1 | NP_001306775.1 | - | - | - | |
NM_023925.4 | NP_076414.2 | - | - | - | |
NM_032156.4 | NP_115532.3 | - | - | - | |
XM_006719138.3 | XP_006719201.1 | - | - | - | |
XM_006719139.3 | XP_006719202.1 | - | - | - | |
XM_006719140.3 | XP_006719203.1 | - | - | - | |
XM_006719142.3 | XP_006719205.1 | - | - | - | |
XM_006719144.3 | XP_006719207.1 | - | - | - | |
XM_006719145.3 | XP_006719208.1 | - | - | - | |
XM_006719146.3 | XP_006719209.1 | - | - | - | |
XM_006719147.3 | XP_006719210.1 | - | - | - | |
XM_017019851.1 | XP_016875340.1 | - | - | - | |
XM_017019852.1 | XP_016875341.1 | - | - | - | |
XM_017019853.1 | XP_016875342.1 | - | - | - | |
XM_017019854.1 | XP_016875343.1 | - | - | - | |
XM_017019855.1 | XP_016875344.1 | - | - | - | |
XM_017019856.1 | XP_016875345.1 | - | - | - | |
XM_017019857.1 | XP_016875346.1 | - | - | - | |
XM_017019858.1 | XP_016875347.1 | - | - | - | |
XM_017019859.1 | XP_016875348.1 | - | - | - | |
XM_017019860.1 | XP_016875349.1 | - | - | - | |
XM_017019861.1 | XP_016875350.1 | - | - | - | |
XM_017019862.1 | XP_016875351.1 | - | - | - | |
XM_017019863.1 | XP_016875352.1 | - | - | - | |
XM_017019865.1 | XP_016875354.1 | - | - | - | |
XM_017019866.1 | XP_016875355.1 | - | - | - | |
XM_017019867.1 | XP_016875356.1 | - | - | - | |
XM_017019868.1 | XP_016875357.1 | - | - | - | |
XM_017019869.1 | XP_016875358.1 | - | - | - | |
XM_017019870.1 | XP_016875359.1 | - | - | - | |
XM_017019871.1 | XP_016875360.1 | - | - | - | |
XM_017019872.1 | XP_016875361.1 | - | - | - | |
XM_017019873.1 | XP_016875362.1 | - | - | - | |
XM_017019874.1 | XP_016875363.1 | - | - | - | |
XM_017019875.1 | XP_016875364.1 | - | - | - | |
XM_017019876.1 | XP_016875365.1 | - | - | - | |
XM_017019877.1 | XP_016875366.1 | - | - | - | |
XM_017019878.1 | XP_016875367.1 | - | - | - | |
XM_017019879.1 | XP_016875368.1 | - | - | - | |
XM_017019880.1 | XP_016875369.1 | - | - | - | |
XM_017019881.1 | XP_016875370.1 | - | - | - | |
XM_017019882.1 | XP_016875371.1 | - | - | - | |
XM_017019883.1 | XP_016875372.1 | - | - | - | |
GenBank | AF525416.1 | AAP80792.1 | - | - | - |
AK024247.1 | - | - | - | ||
AK026222.1 | BAB15398.1 | - | - | - | |
AL834212.1 | - | - | - | ||
AY074490.1 | AAL71549.1 | - | - | - | |
AY074491.1 | AAL71550.1 | - | - | - | |
BC007520.1 | - | - | - | ||
BC025420.2 | - | - | - | ||
BC066295.1 | AAH66295.1 | - | - | - | |
BC111007.1 | AAI11008.1 | - | - | - | |
BQ010043.1 | - | - | - | ||
BX537569.1 | CAD97786.1 | - | - | - | |
BX538080.1 | CAD98004.1 | - | - | - |
Literature Links: |
CAPRIN2 PubMed Links |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
Set Membership: |
Between 200-400 bp |