Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene Symbol:
GPRIN2
LOC107984026
Entrez Gene ID
9721
107984026
Gene Name:
G protein regulated inducer of neurite outgrowth 2
arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
Gene Aliases
GRIN2, KIAA0514
-
Gene Chromosome Location
Chr.10: on Build GRCh38
Chr.10: on Build GRCh38
UniGene
Hs.523375
Species:
Human
Primer Chromosome Location
Chr.10: 46549598-46549349 on Build GRCh38
View in UCSC browser
Amplicon Length
250
Primer Sequences
Log in to view
Ampliseq ID's
Primer ID
Hs00784662_CE
Primer 5'-tail Option
Non-tailed
M13-tailed
Purification
Desalted
HPLC
Primer Pair /FWD/REV
Pair
Forward
Reverse
Synthesis Scale
25 nmol
Availability
Made To Order
Catalog #'s
A15629,A15630
Price
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Information
GPRIN2
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
NM_014696.3
NP_055511.2
4014310
c.1182C>T
p.Y394Y
255923
c.1336C>T
p.R446C
4894477
c.1330C>T
p.L444L
3751795
c.1308A>G
p.P436P
4144708
c.1199T>C
p.L400P
6501482
c.1281C>T
p.S427S
3747186
c.1266C>T
p.P422P
3438024
c.1153G>A
p.D385N
6129665
c.1210G>A
p.G404S
4647183
c.1282C>T
p.L428L
1128052
c.1337G>A
p.R446H
1235914
c.1307C>T
p.P436L
1958326
c.1183G>A
p.G395R
6210501
c.1176G>A
p.E392E
1958322
c.1147C>T
p.R383*
3438025
c.1286C>T
p.S429F
6841541
c.1298G>A
p.R433Q
6837680
c.1349G>T
p.C450F
6852126
c.1365G>A
p.A455A
4392385
c.1329C>T
p.S443S
5370663
c.1194G>A
p.V398V
3751796
c.1359C>T
p.S453S
918371
c.1332G>A
p.L444L
4432432
c.1312C>T
p.R438W
918370
c.1206G>A
p.V402V
5482837
c.1191G>A
p.A397A
5458130
c.1181A>G
p.Y394C
XM_005277666.3
XP_005277723.1
-
-
-
XM_011540388.2
XP_011538690.1
-
-
-
XM_011540395.2
XP_011538697.1
-
-
-
XM_011540400.2
XP_011538702.1
-
-
-
XM_017016974.1
XP_016872463.1
-
-
-
XM_017016975.1
XP_016872464.1
-
-
-
XM_017016976.1
XP_016872465.1
-
-
-
XM_017016977.1
XP_016872466.1
-
-
-
XM_017016978.1
XP_016872467.1
-
-
-
XM_017016979.1
XP_016872468.1
-
-
-
XM_017016980.1
XP_016872469.1
-
-
-
XM_017016981.1
XP_016872470.1
-
-
-
XM_017016982.1
XP_016872471.1
-
-
-
XM_017016983.1
XP_016872472.1
-
-
-
XM_017016984.1
XP_016872473.1
-
-
-
GenBank
AB011086.1
BAA25440.2
-
-
-
BC011672.2
AAH11672.1
-
-
-
ENST00000374317.0
-
-
-
Phenotype
MIM: 611240
LOC107984026
Accession
Protein
COSMIC ID
Mutation
AA
RefSeq
XM_017017029.1
XP_016872518.1
-
-
-
XM_017017030.1
XP_016872519.1
-
-
-
XM_017017031.1
XP_016872520.1
-
-
-
XM_017017032.1
XP_016872521.1
-
-
-
XM_017017033.1
XP_016872522.1
-
-
-
XM_017017034.1
XP_016872523.1
-
-
-
XM_017017038.1
XP_016872527.1
-
-
-
XM_017017039.1
XP_016872528.1
-
-
-
XM_017017040.1
XP_016872529.1
-
-
-
Phenotype
Literature Links:
GPRIN2
PubMed Links
SNP ID
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Process(es)
protein binding
Format:
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer
™
Tool Quick Reference
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0