Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55063 |
Gene Name: | zinc finger CW-type and PWWP domain containing 1 |
Gene Aliases: |
ZCW1 |
Location: |
Chr.7:100400826-100429239 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZCWPW1 | NM_001258008.1 | NP_001244937.1 | ||
NM_017984.4 | NP_060454.3 | |||
XM_005250479.2 | XP_005250536.1 | |||
XM_005250480.2 | XP_005250537.1 | |||
XM_006716035.3 | XP_006716098.1 | |||
XM_006716036.2 | XP_006716099.1 | |||
XM_006716037.3 | XP_006716100.1 | |||
XM_006716038.3 | XP_006716101.1 | |||
XM_006716040.3 | XP_006716103.1 | |||
XM_017012379.1 | XP_016867868.1 | |||
XM_017012380.1 | XP_016867869.1 | |||
AK000919.1 | BAA91424.1 | |||
AK093038.1 | BAC04028.1 | |||
AK300745.1 | ||||
AK302108.1 | ||||
AL136735.1 | CAB66669.1 | |||
BC002725.1 | AAH02725.1 | |||
KU178677.1 | ||||
KU178678.1 | ||||
KU178679.1 | ||||
KU178680.1 | ||||
KU178681.1 | ||||
KU178682.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | MIR4653 MIR93 GIGYF1 GPC2 ZKSCAN1 GNB2 TSC22D4 PLOD3 SRRT MUC3A C7orf43 MIR4658 PVRIG2P PVRIG ZCWPW1 FIS1 MUC17 AP4M1 PCOLCE-AS1 TFR2 SLC12A9 MBLAC1 PILRA UFSP1 EPO TRIP6 SERPINE1 MOGAT3 SAP25 EPHB4 COPS6 STAG3L5P-PVRIG2P-PILRB GAL3ST4 ZAN AZGP1 ZSCAN21 FBXO24 C7orf61 TAF6 NYAP1 ACHE GATS VGF MIR6875 LRCH4 MIR106B ZNHIT1 LOC105375429 STAG3 AP1S1 MIR25 PMS2P1 MIR6840 MCM7 AGFG2 LOC102724094 ZASP MEPCE PPP1R35 PCOLCE MOSPD3 MUC12 STAG3L5P PILRB ACTL6B LAMTOR4 TRIM56 NAT16 CNPY4 POP7 AZGP1P1 SPDYE3 ZNF3 CLDN15 |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | C7orf61 NYAP1 MEPCE PILRA PPP1R35 STAG3L5P TSC22D4 PILRB STAG3L5P-PVRIG2P-PILRB PVRIG2P PMS2P1 SPDYE3 ZCWPW1 MIR6840 |
nsv819923 | Chr.7:100409307 - 100409469 on Build GRCh38 | Loss | ZCWPW1 |
Set Membership: |
Intragenic Non-exonic DGV Variation |