Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23400 |
Gene Name: | ATPase 13A2 |
Gene Aliases: |
CLN12, HSA9947, KRPPD, PARK9 |
Location: |
Chr.1:16985958-17011972 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP13A2 | NM_001141973.2 | 22 | 2676 | NP_001135445.1 |
NM_001141974.2 | NP_001135446.1 | |||
NM_022089.3 | 22 | 2691 | NP_071372.1 | |
XM_005245810.1 | 22 | 2688 | XP_005245867.1 | |
XM_005245811.1 | 22 | 2676 | XP_005245868.1 | |
XM_005245812.1 | 22 | 2664 | XP_005245869.1 | |
XM_005245815.1 | XP_005245872.1 | |||
XM_006710512.1 | 22 | 2673 | XP_006710575.1 | |
XM_006710513.1 | 22 | 2649 | XP_006710576.1 | |
XM_011541128.1 | 22 | 2676 | XP_011539430.1 | |
XM_011541129.1 | 21 | 2484 | XP_011539431.1 | |
XM_017000844.1 | 22 | 2676 | XP_016856333.1 | |
XM_017000845.1 | 22 | 2673 | XP_016856334.1 | |
XM_017000846.1 | 22 | 2649 | XP_016856335.1 | |
XM_017000847.1 | 22 | 2646 | XP_016856336.1 | |
XM_017000848.1 | XP_016856337.1 | |||
XM_017000849.1 | XP_016856338.1 | |||
XM_017000850.1 | 21 | 2484 | XP_016856339.1 | |
AK025928.1 | 2 | 80 | ||
AK075310.1 | 12 | 1473 | BAC11539.1 | |
AK290210.1 | 22 | 2632 | ||
AL354615.1 | 22 | 2491 | CAB89728.1 | |
AL833966.1 | 3 | 345 | CAD38813.2 | |
AY461712.1 | 22 | 2490 | AAR23423.1 | |
BC030267.1 | AAH30267.1 | |||
BC034575.1 | 19 | 2182 | AAH34575.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508947 | Chr.1:16909817 - 17011115 on Build GRCh38 | Insertion | ATP13A2 MFAP2 CROCC |
nsv954854 | Chr.1:16961106 - 17013905 on Build GRCh38 | Deletion | ATP13A2 MFAP2 CROCC |
nsv428421 | Chr.1:16437838 - 17157486 on Build GRCh38 | Gain+Loss | ESPNP FAM231A NBPF1 MIR3675 RNU1-3 LOC105376796 LOC100132147 SPATA21 RNU1-1 MST1P2 LOC102723383 FAM231B CROCC LOC105376805 CROCCP3 MST1L PADI2 NECAP2 CROCCP2 SDHB RNU1-4 RNU1-2 ATP13A2 MFAP2 |
dgv108n100 | Chr.1:16850538 - 17010370 on Build GRCh38 | Gain | MIR3675 RNU1-2 ATP13A2 MFAP2 CROCC LOC105376805 |
dgv92n100 | Chr.1:16684943 - 17003642 on Build GRCh38 | Gain | ESPNP MIR3675 LOC102723383 RNU1-4 RNU1-2 ATP13A2 MFAP2 CROCC LOC105376805 MST1L |
nsv834280 | Chr.1:16888218 - 17123006 on Build GRCh38 | Loss | PADI2 SDHB RNU1-2 ATP13A2 MFAP2 CROCC LOC105376805 |
dgv120n100 | Chr.1:16890896 - 16997938 on Build GRCh38 | Gain | RNU1-2 ATP13A2 MFAP2 CROCC |
nsv834269 | Chr.1:16855283 - 17038531 on Build GRCh38 | Loss | MIR3675 SDHB RNU1-2 ATP13A2 MFAP2 CROCC LOC105376805 |
Set Membership: |
Intragenic Exonic DGV Variation |