Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5189 |
Gene Name: | peroxisomal biogenesis factor 1 |
Gene Aliases: |
HMLR1, PBD1A, PBD1B, ZWS, ZWS1 |
Location: |
Chr.7:92487023-92528531 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PEX1 | NM_000466.2 | NP_000457.1 | ||
NM_001282677.1 | NP_001269606.1 | |||
NM_001282678.1 | NP_001269607.1 | |||
XM_017012319.1 | XP_016867808.1 | |||
AB008112.1 | BAA85162.1 | |||
AB052090.1 | BAB59061.1 | |||
AB052091.1 | BAB59062.1 | |||
AB052092.1 | BAB59063.1 | |||
AB052093.1 | BAB59064.1 | |||
AB052094.1 | BAB59065.1 | |||
AB209355.1 | 1 | 787 | BAD92592.1 | |
AF026086.1 | AAB87880.1 | |||
AF030356.1 | AAB99758.1 | |||
AK292955.1 | ||||
AK293757.1 | ||||
AK293848.1 | ||||
AK295686.1 | ||||
BC035575.1 | AAH35575.1 |
Set Membership: |
Intragenic Non-exonic non-DGV Variation |