Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 50943 |
Gene Name: | forkhead box P3 |
Gene Aliases: |
AIID, DIETER, IPEX, JM2, PIDX, XPID |
Location: |
Chr.X:49250436-49266505 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXP3 | NM_001114377.1 | 1 | 127 | NP_001107849.1 |
NM_014009.3 | 1 | 127 | NP_054728.2 | |
XM_006724533.2 | 1 | 233 | XP_006724596.2 | |
XM_011543916.2 | 2 | 1657 | XP_011542218.1 | |
XM_017029565.1 | 2 | 1657 | XP_016885054.1 | |
XM_017029566.1 | 2 | 1657 | XP_016885055.1 | |
AF277993.1 | 1 | 127 | AAG53607.1 | |
AK292052.1 | ||||
AK299988.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain | GAGE10 PPP1R3F CCDC22 FOXP3 CACNA1F |
nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain | KCND1 HDAC6 PRAF2 GATA1 CCDC120 WDR45 WAS WDR13 GPKOW RBM3 ERAS SLC38A5 MAGIX PORCN PRICKLE3 GRIPAP1 SYP-AS1 PLP2 LOC101927635 GLOD5 CACNA1F SUV39H1 PCSK1N SYP CCDC22 TBC1D25 FOXP3 TIMM17B SLC35A2 PIM2 OTUD5 TFE3 PPP1R3F EBP PQBP1 FTSJ1 |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss | KCND1 HDAC6 PRAF2 SSX4B GATA1 ZNF630-AS1 CCDC120 WDR45 WAS ZNF182 WDR13 GPKOW RBM3 ERAS SLC38A5 MAGIX SSX9 PORCN PRICKLE3 GRIPAP1 SYP-AS1 SPACA5B PLP2 LOC101927635 GLOD5 CACNA1F SUV39H1 SSX1 PCSK1N SSX3 SYP CCDC22 SSX4 TBC1D25 FOXP3 TIMM17B SLC35A2 ZNF630 PIM2 SPACA5 OTUD5 TFE3 PPP1R3F SSX6 EBP PQBP1 FTSJ1 SSX5 |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss | GAGE10 PPP1R3F CCDC22 FOXP3 CACNA1F |
Set Membership: |
Intragenic Exonic DGV Variation |