Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 55063 |
Gene Name: | zinc finger CW-type and PWWP domain containing 1 |
Gene Aliases: |
ZCW1 |
Location: |
Chr.7:100400826-100429239 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZCWPW1 | NM_001258008.1 | NP_001244937.1 | ||
NM_017984.4 | NP_060454.3 | |||
XM_005250479.2 | XP_005250536.1 | |||
XM_005250480.2 | XP_005250537.1 | |||
XM_006716035.3 | XP_006716098.1 | |||
XM_006716036.2 | XP_006716099.1 | |||
XM_006716037.3 | XP_006716100.1 | |||
XM_006716038.3 | XP_006716101.1 | |||
XM_006716040.3 | XP_006716103.1 | |||
XM_017012379.1 | XP_016867868.1 | |||
XM_017012380.1 | XP_016867869.1 | |||
AK000919.1 | BAA91424.1 | |||
AK093038.1 | BAC04028.1 | |||
AK300745.1 | ||||
AK302108.1 | ||||
AL136735.1 | CAB66669.1 | |||
BC002725.1 | AAH02725.1 | |||
KU178677.1 | ||||
KU178678.1 | ||||
KU178679.1 | ||||
KU178680.1 | ||||
KU178681.1 | ||||
KU178682.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | ZAN SLC12A9 AP4M1 ACHE MUC17 PCOLCE MIR106B ZSCAN21 VGF NAT16 NYAP1 PMS2P1 TSC22D4 CLDN15 ZKSCAN1 PVRIG2P MBLAC1 ZASP UFSP1 C7orf43 GAL3ST4 AP1S1 MIR25 SERPINE1 PCOLCE-AS1 MOSPD3 LRCH4 AZGP1 MUC3A MOGAT3 ZNHIT1 SRRT ZCWPW1 GIGYF1 MIR4658 GPC2 GNB2 COPS6 POP7 PLOD3 ZNF3 PVRIG LOC105375429 LAMTOR4 ACTL6B TFR2 FBXO24 MCM7 AGFG2 FIS1 SPDYE3 C7orf61 EPHB4 MEPCE TRIM56 PPP1R35 EPO LOC102724094 AZGP1P1 TAF6 TRIP6 PILRA MIR6840 CNPY4 STAG3L5P MIR93 GATS MIR6875 STAG3L5P-PVRIG2P-PILRB SAP25 MIR4653 STAG3 MUC12 PILRB |
nsv831071 | Chr.7:100320402 - 100515790 on Build GRCh38 | Loss | PILRA MIR6840 STAG3L5P NYAP1 PMS2P1 SPDYE3 C7orf61 MEPCE TSC22D4 STAG3L5P-PVRIG2P-PILRB PPP1R35 ZCWPW1 PVRIG2P PILRB |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |