Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 2692 |
Gene Name: | growth hormone releasing hormone receptor |
Gene Aliases: |
GHRFR, GRFR, IGHD1B |
Location: |
Chr.7:30964021-30979531 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GHRHR | NM_000823.3 | NP_000814.2 | ||
AY557192.1 | AAS59864.1 | |||
L01406.1 | AAA35890.1 | |||
L09237.1 | AAA58619.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv6316n100 | Chr.7:30609596 - 31145482 on Build GRCh38 | Gain | GHRHR INMT AQP1 CRHR2 INMT-FAM188B ADCYAP1R1 FAM188B GARS |
nsv527038 | Chr.7:30617851 - 31122331 on Build GRCh38 | Gain | GHRHR INMT AQP1 CRHR2 INMT-FAM188B ADCYAP1R1 FAM188B GARS |
esv2761316 | Chr.7:30615466 - 31127062 on Build GRCh38 | Gain | GHRHR INMT AQP1 CRHR2 INMT-FAM188B ADCYAP1R1 FAM188B GARS |
nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion | LINC01176 LOC101928268 GGCT DKFZP586I1420 INMT-FAM188B ZNRF2P1 ADCYAP1R1 LOC401320 NOD1 AVL9 GHRHR DPY19L1P1 NEUROD6 AQP1 CCDC129 CRHR2 FAM188B MIR550B1 INMT PPP1R17 LOC100130673 ZNRF2 MIR550A1 PDE1C LSM5 GARS |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |