Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23230 |
Gene Name: | vacuolar protein sorting 13 homolog A |
Gene Aliases: |
CHAC, CHOREIN |
Location: |
Chr.9:77177353-77421537 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VPS13A | NM_001018037.1 | NP_001018047.1 | ||
NM_001018038.2 | NP_001018048.1 | |||
NM_015186.3 | NP_056001.1 | |||
NM_033305.2 | NP_150648.2 | |||
AB054005.1 | BAB59128.1 | |||
AF337532.1 | AAK61861.1 | |||
AJ608769.1 | CAE75581.1 | |||
AJ626859.1 | CAF25186.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3620768 | Chr.9:77186688 - 77228922 on Build GRCh38 | Loss | VPS13A |
nsv1041647 | Chr.9:77211923 - 77537136 on Build GRCh38 | Gain | GNA14-AS1 VPS13A GNA14 |
nsv614728 | Chr.9:77195973 - 77375037 on Build GRCh38 | Loss | VPS13A |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |