Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23624 |
Gene Name: | Cbl proto-oncogene C |
Gene Aliases: |
CBL-3, CBL-SL, RNF57 |
Location: |
Chr.19:44777855-44800646 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CBLC | NM_001130852.1 | NP_001124324.1 | ||
NM_012116.3 | NP_036248.3 | |||
XM_011526688.2 | XP_011524990.1 | |||
XM_011526689.2 | XP_011524991.1 | |||
AB028645.1 | BAA86298.1 | |||
AF117646.1 | AAD34341.1 | |||
AF117647.1 | AAD34342.1 | |||
BC028915.1 | AAH28915.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833841 | Chr.19:44705217 - 44804786 on Build GRCh38 | Loss | BCL3 CEACAM16 CBLC MIR8085 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |