Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 9211 |
Gene Name: | leucine rich glioma inactivated 1 |
Gene Aliases: |
ADLTE, ADPAEF, ADPEAF, EPITEMPIN, EPT, ETL1, IB1099 |
Location: |
Chr.10:93757770-93798174 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LGI1 | NM_001308275.1 | NP_001295204.1 | ||
NM_001308276.1 | NP_001295205.1 | |||
NM_005097.3 | NP_005088.1 | |||
NR_131777.1 | ||||
XM_017016911.1 | XP_016872400.1 | |||
XM_017016912.1 | XP_016872401.1 | |||
AF055636.1 | AAC99316.1 | |||
AF473548.1 | AAM22074.1 | |||
AK289706.1 | ||||
AK303956.1 | ||||
AK309416.1 | ||||
AY358885.1 | AAQ89244.1 | |||
BC022500.1 | AAH22500.1 | |||
DC326684.1 | ||||
KU178468.1 | ||||
KU178469.1 | ||||
KU178470.1 | ||||
KU178471.1 |
Set Membership: |
Intragenic Non-exonic non-DGV Variation |