Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 3636 |
Gene Name: | inositol polyphosphate phosphatase like 1 |
Gene Aliases: |
OPSMD, SHIP2 |
Location: |
Chr.11:72223861-72239147 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
INPPL1 | NM_001567.3 | 28 | 4083 | NP_001558.3 |
XM_005273978.3 | 29 | 4460 | XP_005274035.1 | |
XM_005273979.4 | 29 | 4307 | XP_005274036.1 | |
XM_011544999.2 | 29 | 4241 | XP_011543301.1 | |
XM_011545000.2 | 28 | 4173 | XP_011543302.1 | |
XM_017017673.1 | 30 | 4398 | XP_016873162.1 | |
L24444.1 | 3 | 673 | AAA50503.1 | |
L36818.1 | 27 | 4061 | AAA96658.1 | |
Y14385.1 | 28 | 4147 | CAA74743.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951038 | Chr.11:72226157 - 72253456 on Build GRCh38 | Deletion | PHOX2A INPPL1 LOC220077 |
nsv555405 | Chr.11:72233623 - 72241301 on Build GRCh38 | Loss | PHOX2A INPPL1 |
nsv555404 | Chr.11:72233198 - 72238908 on Build GRCh38 | Gain+Loss | INPPL1 |
Set Membership: |
Intragenic Exonic DGV Variation |