Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 90678 |
Gene Name: | leucine rich repeat and sterile alpha motif containing 1 |
Gene Aliases: |
CMT2P, RIFLE, TAL |
Location: |
Chr.9:127451486-127503501 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRSAM1 | NM_001005373.3 | 26 | 2721 | NP_001005373.1 |
NM_001005374.3 | 25 | 2426 | NP_001005374.1 | |
NM_001190723.2 | 25 | 2640 | NP_001177652.1 | |
NM_138361.5 | 25 | 2980 | NP_612370.3 | |
XM_006717316.3 | 25 | 2622 | XP_006717379.1 | |
XM_017015283.1 | 26 | 3720 | XP_016870772.1 | |
XM_017015284.1 | 26 | 2716 | XP_016870773.1 | |
AK056203.1 | 25 | 2622 | BAB71119.1 | |
AK056305.1 | 14 | 1571 | BAB71144.1 | |
AK091589.1 | 25 | 2728 | BAC03703.1 | |
AK097094.1 | 6 | 2181 | ||
AK226106.1 | 26 | 2707 | ||
AY358830.1 | 26 | 2686 | AAQ89189.1 | |
BC009239.2 | 25 | 2426 | AAH09239.1 | |
DB309230.1 | 1 | 426 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv466569 | Chr.9:127470144 - 127512011 on Build GRCh38 | Loss | LRSAM1 FAM129B |
Set Membership: |
Intragenic Exonic DGV Variation |