Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 79628 |
Gene Name: | SH3 domain and tetratricopeptide repeats 2 |
Gene Aliases: |
CMT4C, MNMN |
Location: |
Chr.5:148982150-149063174 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SH3TC2 | NM_024577.3 | 6 | 811 | NP_078853.2 |
AB075865.1 | BAB85571.1 | |||
AK127241.1 | 1 | 59 | ||
AK127248.1 | BAC86899.1 | |||
AK300420.1 | 7 | 767 | ||
AY341075.1 | 6 | 761 | AAR03497.1 | |
BC113879.1 | 6 | 726 | ||
BC114486.1 | 6 | 705 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830517 | Chr.5:148953659 - 149171578 on Build GRCh38 | Gain | MIR584 LOC255187 ABLIM3 SH3TC2 |
Set Membership: |
Intragenic Exonic DGV Variation |