Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 5071 |
Gene Name: | parkin RBR E3 ubiquitin protein ligase |
Gene Aliases: |
AR-JP, LPRS2, PDJ, PRKN |
Location: |
Chr.6:161347417-162727802 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PARK2 | NM_004562.2 | NP_004553.2 | ||
NM_013987.2 | NP_054642.2 | |||
NM_013988.2 | NP_054643.2 | |||
XM_011535863.1 | XP_011534165.1 | |||
XM_017010908.1 | XP_016866397.1 | |||
XM_017010909.1 | XP_016866398.1 | |||
AB009973.1 | BAA25751.1 | |||
AB245403.1 | ||||
AF381282.1 | AAM21457.1 | |||
AF381283.1 | AAM21458.1 | |||
AF381284.1 | AAM21459.1 | |||
AK292590.1 | ||||
AK294684.1 | ||||
BC022014.2 | AAH22014.1 | |||
EF375726.1 | ||||
GU345837.1 | ||||
GU345838.1 | ||||
GU345839.1 | ||||
GU345840.1 | ||||
GU357501.1 | ||||
GU357502.1 | ||||
GU361466.1 | ||||
GU361467.1 | ||||
GU361468.1 | ||||
GU361469.1 | ||||
GU361470.1 | ||||
GU361471.1 | ||||
KC357595.1 | ||||
KC774171.1 | ||||
KJ534911.1 | ||||
KJ534912.1 | ||||
KU178240.1 | ||||
KU178241.1 | ||||
KU178242.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026147 | Chr.6:161704721 - 162260681 on Build GRCh38 | Loss | PARK2 |
nsv5578 | Chr.6:161707243 - 161765549 on Build GRCh38 | Insertion | PARK2 |
dgv6169n100 | Chr.6:161644335 - 161956796 on Build GRCh38 | Loss | PARK2 |
esv3611467 | Chr.6:161745455 - 161836350 on Build GRCh38 | Gain | PARK2 |
esv3611465 | Chr.6:161567774 - 162085535 on Build GRCh38 | Gain | PARK2 |
esv2422275 | Chr.6:161628725 - 161781763 on Build GRCh38 | Duplication | PARK2 |
nsv1031234 | Chr.6:161255736 - 161829694 on Build GRCh38 | Gain | LOC105378098 PARK2 AGPAT4 |
esv3611469 | Chr.6:161760442 - 161774964 on Build GRCh38 | Gain | PARK2 |
nsv1029749 | Chr.6:161543443 - 162045027 on Build GRCh38 | Loss | PARK2 |
nsv518931 | Chr.6:161372287 - 161985303 on Build GRCh38 | Gain | LOC105378098 PARK2 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |