Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 11315 |
Gene Name: | Parkinsonism associated deglycase |
Gene Aliases: |
DJ-1, DJ1, HEL-S-67p |
Location: |
Chr.1:7961654-7985282 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PARK7 | NM_001123377.1 | NP_001116849.1 | ||
NM_007262.4 | NP_009193.2 | |||
XM_005263424.3 | XP_005263481.1 | |||
AB073864.1 | BAB71782.1 | |||
AF021819.1 | AAC12806.1 | |||
AK130886.1 | ||||
AK312000.1 | ||||
BC008188.2 | AAH08188.1 | |||
BP248330.1 | ||||
D61380.2 | BAA09603.2 | |||
EU794641.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1521 | Chr.1:7954179 - 7987433 on Build GRCh38 | Insertion | PARK7 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |