Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 23500 |
Gene Name: | dishevelled associated activator of morphogenesis 2 |
Gene Aliases: |
dJ90A20A.1 |
Location: |
Chr.6:39792366-39904877 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DAAM2 | NM_001201427.1 | NP_001188356.1 | ||
NM_015345.3 | NP_056160.2 | |||
XM_006715039.3 | XP_006715102.3 | |||
XM_006715040.3 | XP_006715103.3 | |||
XM_006715042.2 | XP_006715105.1 | |||
XM_006715043.2 | XP_006715106.1 | |||
XM_006715045.3 | XP_006715108.1 | |||
XM_006715046.3 | XP_006715109.1 | |||
XM_017010630.1 | XP_016866119.1 | |||
AB002379.2 | ||||
AK310941.1 | ||||
AL833083.2 | CAD89973.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv462921 | Chr.6:39861447 - 39891891 on Build GRCh38 | Loss | LOC100505635 DAAM2 |
Set Membership: |
Intragenic Non-exonic DGV Variation |