Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 73750 |
Gene Name: | whirlin |
Gene Aliases: |
1110035G07Rik, AW122018, AW742671, C430046P22Rik, Dfnb31, wi |
Location: |
Chr.4:63414909-63496078 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Whrn | NM_001008791.2 | 10 | 2932 | NP_001008791.1 |
NM_001008792.2 | 9 | 2911 | NP_001008792.1 | |
NM_001008793.2 | 9 | 2899 | NP_001008793.1 | |
NM_028640.2 | 9 | 2899 | NP_082916.1 | |
AK122523.1 | 9 | 2030 | BAC65805.1 | |
AY739114.1 | 9 | 2859 | AAV87519.1 | |
AY739115.1 | 10 | 2892 | AAV87520.1 | |
AY739116.1 | 9 | 2871 | AAV87521.1 | |
AY739117.1 | 9 | 2859 | AAV87522.1 | |
AY739118.1 | 6 | 1508 | AAV87523.1 | |
AY739119.1 | 5 | 1475 | AAV87524.1 | |
AY739120.1 | 4 | 1262 | AAV87525.1 | |
AY739121.1 | 4 | 1380 | AAV87526.1 | |
AY739122.1 | 5 | 1712 | AAV87527.1 | |
BC150757.1 | 9 | 2256 |
Set Membership: |
Intragenic Exonic |