Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 66725 |
Gene Name: | leucine-rich repeat kinase 2 |
Gene Aliases: |
4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46 |
Location: |
Chr.15:91672821-91816124 on Build GRCm38 |
Assay Gene Location: | Within Exon 51 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrrk2 | NM_025730.3 | 51 | 8104 | NP_080006.3 |
AK014886.1 | BAB29603.1 | |||
AK050702.1 | 2 | 691 | ||
AK156100.1 | ||||
BC034074.1 | AAH34074.1 | |||
BC035949.1 | AAH35949.1 | |||
BF660966.1 | 1 | 128 |
Set Membership: |
Intragenic Exonic |