Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CXCL12 EBGEX Assays ›
          Gene Symbol:
          CXCL12
          Gene Name:
          C-X-C motif chemokine ligand 12
          Gene Aliases:
          IRH, PBSF, SCYB12, SDF1, TLSF, TPAR1
          Chromosome Location:
          Chr.10: 44370165 - 44385097 on Build GRCh38
          UniGene ID:
          Hs.522891
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Type:
          1
          Detection Label:
          Alexa Fluor 647
          ProductType:
          PrimeFlow Probe Set
          Assay ID VA1-11035-PF
          Size
          Availability Made To Order
          Catalog # PF-204
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CXCL12
          Interrogated Sequence Translated Protein IMAGE Clone ID
          RefSeq NM_000609.7 NP_000600.1
          NM_001033886.2 NP_001029058.1
          NM_001178134.2 NP_001171605.1
          NM_001277990.2 NP_001264919.1
          NM_199168.4 NP_954637.1
          GenBank mRNA AI092156.1 -
          AJ227905.1 -
          AK090482.1 -
          AK124641.1 -
          AK292628.1 -
          AK311814.1 -
          AL713778.1 -
          AU120056.1 -
          AW014388.1 -
          AY644456.1 -
          AY874118.1 -
          BC031072.1 -
          BC039893.1 -
          BX647204.1 -
          CD172464.1 -
          CR450283.1 -
          DA266752.1 -
          DQ345517.1 -
          DQ345518.1 -
          DQ345519.1 -
          DQ345520.1 -
          L36033.1 -
          L36034.1 -
          U16752.1 -

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neuron migration
          positive regulation of endothelial cell proliferation
          cellular calcium ion homeostasis
          chemotaxis
          defense response
          immune response
          cell adhesion
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          axon guidance
          blood circulation
          regulation of actin polymerization or depolymerization
          positive regulation of cell proliferation
          adult locomotory behavior
          response to mechanical stimulus
          response to virus
          T cell chemotaxis
          telencephalon cell migration
          positive regulation of cell migration
          organ regeneration
          killing of cells of other organism
          positive regulation of dopamine secretion
          integrin activation
          chemokine (C-X-C motif) ligand 12 signaling pathway
          CXCL12-activated CXCR4 signaling pathway
          response to peptide hormone
          positive regulation of neuron differentiation
          positive regulation of cell adhesion
          positive regulation of axon extension involved in axon guidance
          positive regulation of chemotaxis
          induction of positive chemotaxis
          detection of temperature stimulus involved in sensory perception of pain
          detection of mechanical stimulus involved in sensory perception of pain
          regulation of calcium ion transport
          cell chemotaxis
          antimicrobial humoral immune response mediated by antimicrobial peptide
          chemokine-mediated signaling pathway
          positive regulation of monocyte chemotaxis
          positive regulation of calcium ion import
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of leukocyte tethering or rolling
          positive regulation of vasculature development
          response to ultrasound
          cellular response to chemokine
          positive regulation of T cell migration
          negative regulation of dendritic cell apoptotic process
          receptor binding
          cytokine activity
          integrin binding
          protein binding
          chemokine activity
          growth factor activity
          chemokine receptor binding
          CXCR chemokine receptor binding

          Back To Top
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline