Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKFY1 EBGEX Assays ›
          Gene Symbol:
          ANKFY1
          Gene Name:
          ankyrin repeat and FYVE domain containing 1
          Gene Aliases:
          ANKHZN, BTBD23, ZFYVE14
          Chromosome Location:
          Chr.17: 4163821 - 4263979 on Build GRCh38
          UniGene ID:
          Hs.696087
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Type:
          1
          Detection Label:
          Alexa Fluor 647
          ProductType:
          PrimeFlow Probe Set
          Assay ID VA1-3009038-PF
          Size 100 tests
          Availability Made To Order
          Catalog # PF-210
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANKFY1
          Interrogated Sequence Translated Protein IMAGE Clone ID
          RefSeq NM_001257999.3 NP_001244928.1
          NM_001330063.2 NP_001316992.1
          NM_016376.5 NP_057460.3
          NR_047571.3
          XM_011523926.2 XP_011522228.1
          XM_017024733.2 XP_016880222.1
          XM_047436221.1 XP_047292177.1
          XM_047436223.1 XP_047292179.1
          XM_047436224.1 XP_047292180.1
          XM_047436225.1 XP_047292181.1
          GenBank mRNA AA897044.1 -
          AB033081.2 -
          AB037360.1 -
          AB073897.1 -
          AK025483.1 -
          AK025717.1 -
          AK025960.1 -
          AK057047.1 -
          AK074324.1 -
          AK091008.1 -
          AK225170.1 -
          AK292930.1 -
          AK302712.1 -
          AL706226.1 -
          BC052308.1 -
          BC060812.1 -
          CR933717.1 -
          DA856905.1 -

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          endosomal transport
          endosomal vesicle fusion
          retrograde transport, endosome to Golgi
          positive regulation of pinocytosis
          Golgi to lysosome transport
          protein binding
          zinc ion binding
          small GTPase binding
          metal ion binding
          phosphatidylinositol phosphate binding

          Back To Top
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline