Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNM1 EBGEX Assays ›
          Gene Symbol:
          Dnm1
          Gene Name:
          dynamin 1
          Gene Aliases:
          AI838169, Dnm, Ftfl, mKIAA4093
          Chromosome Location:
          Chr.2: 32308471 - 32353339 on Build GRCm38
          UniGene ID:
          Mm.44736
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Type:
          1
          Detection Label:
          Alexa Fluor 546
          ProductType:
          ViewRNA Cell Plus Probe Set
          Assay ID VB1-3029504-VCP
          Size
          Availability Made To Order
          Catalog # VX-01
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnm1
          Interrogated Sequence Translated Protein IMAGE Clone ID
          RefSeq NM_001301737.1 NP_001288666.1
          NM_010065.3 NP_034195.2
          NR_125959.1
          GenBank mRNA AA387428.1 -
          AI894003.1 -
          AK011651.1 -
          AK144142.1 -
          AK220483.1 -
          BC034679.1 - 4506865
          BC058623.1 - 5706803
          BE860433.1 -
          BM950315.1 -
          BQ179370.1 -
          BY297150.1 -
          CA328097.1 -
          CO425806.1 -
          CX239206.1 -
          L29457.1 -
          L31395.1 -
          L31396.1 -
          L31397.1 -

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor internalization
          endocytosis
          receptor-mediated endocytosis
          endosome organization
          sensory perception of sound
          adult locomotory behavior
          receptor internalization
          protein tetramerization
          synaptic transmission, GABAergic
          clathrin-mediated endocytosis
          toxin transport
          positive regulation of synaptic vesicle recycling
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          protein C-terminus binding
          hydrolase activity
          protein kinase binding
          D2 dopamine receptor binding
          protein complex binding
          identical protein binding
          poly(A) RNA binding
          protein dimerization activity
          nitric-oxide synthase binding

          Back To Top
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sk2zd:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0