Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CEACAM1 EBGEX Assays ›
          Gene Symbol:
          Ceacam1
          Gene Name:
          carcinoembryonic antigen-related cell adhesion molecule 1
          Gene Aliases:
          Bgp, Bgp1, C-CAM, CD66a, Cc1, Cea-1, Cea-7, Cea1, Cea7, Hv-2, Hv2, MHVR, MHVR1, Mhv-1, bb-1, mCEA1, mmCGM1, mmCGM1a, mmCGM2
          Chromosome Location:
          Chr.7: 25461702 - 25477625 on Build GRCm38
          UniGene ID:
          Mm.322502
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Type:
          6
          Detection Label:
          FastBlue
          ProductType:
          ViewRNA Tissue Probe Set
          Assay ID VB6-3202593-VT
          Size
          Availability Made To Order
          Catalog # VX-01
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ceacam1
          Interrogated Sequence Translated Protein IMAGE Clone ID
          RefSeq NM_001039185.1 NP_001034274.1
          NM_001039186.1 NP_001034275.1
          NM_001039187.1 NP_001034276.1
          NM_011926.2 NP_036056.2
          GenBank mRNA AB236328.2 -
          AB236329.1 -
          AB236330.2 -
          AB236331.2 -
          AB236332.2 -
          AK085408.1 -
          AK085749.1 -
          BC016891.1 -
          BC114998.1 -
          BC121189.1 -
          M61907.1 -
          M77196.1 -
          M96934.1 -
          M96935.1 -
          X15351.1 -
          X53084.1 -
          X67278.1 -
          X67279.1 -
          X67280.1 -
          X67281.1 -
          X67282.1 -
          X67283.1 -

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cytokine production
          homophilic cell adhesion via plasma membrane adhesion molecules
          viral process
          calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          negative regulation of interleukin-2 production
          positive regulation of T cell proliferation
          negative regulation of T cell proliferation
          positive regulation of MAP kinase activity
          negative regulation by host of viral process
          positive regulation by host of viral process
          negative regulation by host of viral genome replication
          negative regulation of JNK cascade
          positive regulation of JNK cascade
          viral entry into host cell
          Peyer's patch development
          positive regulation of immunoglobulin secretion
          positive regulation of NFAT protein import into nucleus
          positive regulation of activation-induced cell death of T cells
          positive regulation of CD4-positive, alpha-beta T cell activation
          positive regulation of CD4-positive, alpha-beta T cell proliferation
          positive regulation of CD8-positive, alpha-beta T cell activation
          virus receptor activity
          protein binding
          protein kinase binding
          protein homodimerization activity
          virion binding

          Back To Top
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-bxd7r:80/100.66.79.78:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline