Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLA1 EBGEX Assays ›
Gene Symbol:
CLA1
Gene Name:
Deoxyxylulose-5-phosphate synthase
Gene Aliases:
1-DEOXY-D-XYLULOSE 5-PHOSPHATE SYNTHASE, 1-DEOXY-D-XYLULOSE 5-PHOSPHATE SYNTHASE 2, 1-deoxy-D-xylulose 5-phosphate (DXP) synthase 1, AT4G15560, AtCLA1, CLA, CLOROPLASTOS ALTERADOS 1, DEF, DL3821W, DXPS2, DXS, DXS1, FCAALL.24
Chromosome Location:
Chr.4: 8883825 - 8887667 on Build TAIR10.1
UniGene ID:
At.23240
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Type:
6
Detection Label:
FastBlue
ProductType:
ViewRNA Tissue Probe Set
Assay ID
VF6-4025938-VT
Size
M: 440 uL
L: 1.44 mL
Availability
Made To Order
Catalog #
VX-01
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CLA1
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_117647.3
NP_193291.1
GenBank mRNA
BT002340.1
-
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
chlorophyll biosynthetic process
terpenoid biosynthetic process
isopentenyl diphosphate biosynthetic process, methylerythritol 4-phosphate pathway
1-deoxy-D-xylulose 5-phosphate biosynthetic process
protein binding
1-deoxy-D-xylulose-5-phosphate synthase activity
transferase activity, transferring aldehyde or ketonic groups
identical protein binding
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0