Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101369.2 | NP_172953.1 | 1 | 1326 | 139 | |
GenBank mRNA | AY075612.1 | - | 1 | 1326 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |