Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QSOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332143.1 | NP_001321934.1 | 7 - 8 | 1041 | 91 | |
NM_101371.5 | NP_172955.1 | 7 - 8 | 1091 | 91 | ||
NM_179333.2 | NP_849664.1 | 7 - 8 | 1041 | 91 | ||
GenBank mRNA | AY062528.1 | - | 7 - 8 | 832 | 91 | |
BT001218.1 | - | 7 - 8 | 717 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |