Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101389.3 | NP_172973.1 | 6 - 7 | 2771 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |