Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCMH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101390.2 | NP_563966.1 | 2 - 3 | 697 | 61 | |
NM_202105.2 | NP_973834.1 | 3 - 4 | 576 | 61 | ||
GenBank mRNA | AY050392.1 | - | 2 - 3 | 699 | 61 | |
AY085498.1 | - | 3 - 4 | 556 | 61 | ||
AY097355.1 | - | 2 - 3 | 276 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |