Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCA2;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101426.5 | NP_173010.1 | 10 - 11 | 1647 | 76 | |
GenBank mRNA | AY072105.1 | - | 10 - 11 | 1492 | 76 | |
AY096733.1 | - | 9 - 10 | 1301 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |