Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035973.3 | NP_001031050.1 | 18 - 19 | 3237 | 103 | |
NM_001035974.4 | NP_001031051.1 | 19 - 20 | 2971 | 103 | ||
NM_101443.4 | NP_563981.1 | 19 - 20 | 3131 | 103 | ||
NM_179341.3 | NP_849672.1 | 20 - 21 | 3041 | 103 | ||
GenBank mRNA | AY046013.1 | - | 19 - 20 | 2901 | 103 | |
AY142619.1 | - | 18 - 19 | 2737 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |