Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035973.3 | NP_001031050.1 | 21 - 22 | 3675 | 67 | |
NM_001035974.4 | NP_001031051.1 | 22 - 23 | 3409 | 67 | ||
NM_101443.4 | NP_563981.1 | 22 - 23 | 3569 | 67 | ||
NM_179341.3 | NP_849672.1 | 23 - 24 | 3479 | 67 | ||
GenBank mRNA | AY046013.1 | - | 22 - 23 | 3339 | 67 | |
AY142619.1 | - | 21 - 22 | 3175 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |