Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101456.4 | NP_563985.1 | 1 - 2 | 314 | 68 | |
GenBank mRNA | AF332426.1 | - | 1 - 2 | 44 | 68 | |
AF357528.1 | - | 1 - 2 | 294 | 68 | ||
AF410263.1 | - | 1 - 2 | 286 | 68 | ||
AY093723.1 | - | 1 - 2 | 44 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |