Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRAMP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101464.4 | NP_173048.3 | 10 - 11 | 2079 | 73 | |
GenBank mRNA | AJ291831.1 | - | 10 - 11 | 1040 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |