Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAKL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101478.3 | NP_173062.1 | 2 - 3 | 1240 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |