Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332212.1 | NP_001323115.1 | 2 - 3 | 656 | 88 | |
NM_101486.3 | NP_173070.1 | 3 - 4 | 410 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |