Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332212.1 | NP_001323115.1 | 6 - 7 | 1082 | 66 | |
NM_101486.3 | NP_173070.1 | 7 - 8 | 836 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |