Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCB3;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101499.4 | NP_173083.3 | 22 - 23 | 1877 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |