Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035979.1 | NP_001031056.1 | 9 - 10 | 953 | 68 | |
NM_001198083.1 | NP_001185012.1 | 10 - 11 | 1026 | 68 | ||
NM_001332226.1 | NP_001322506.1 | 9 - 10 | 937 | 68 | ||
NM_001332227.1 | NP_001322507.1 | 9 - 10 | 947 | 68 | ||
NM_101511.6 | NP_563998.1 | 9 - 10 | 997 | 68 | ||
NM_179344.2 | NP_849675.1 | 9 - 10 | 937 | 68 | ||
GenBank mRNA | AB032865.1 | - | 9 - 10 | 859 | 68 | |
AJ010500.1 | - | 9 - 10 | 867 | 68 | ||
AK229282.1 | - | 9 - 10 | 956 | 68 | ||
AY084558.1 | - | 9 - 10 | 937 | 68 | ||
BT024876.1 | - | 8 - 9 | 794 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |