Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035980.1 | NP_001031057.1 | 2 - 3 | 398 | 105 | |
NM_101512.4 | NP_173096.1 | 3 - 4 | 301 | 105 | ||
GenBank mRNA | AF043520.1 | - | 3 - 4 | 234 | 105 | |
AF332467.1 | - | 2 - 3 | 114 | 105 | ||
AK317085.1 | - | 3 - 4 | 247 | 105 | ||
AK317410.1 | - | 2 - 3 | 399 | 105 | ||
AY088628.1 | - | 3 - 4 | 247 | 105 | ||
Y13176.1 | - | 234 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |