Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332231.1 | NP_001322902.1 | 17 - 18 | 2280 | 94 | |
NM_101519.3 | NP_564001.1 | 18 - 19 | 2211 | 94 | ||
GenBank mRNA | AF325457.1 | - | 18 - 19 | 2166 | 94 | |
AY034895.1 | - | 18 - 19 | 2116 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |