Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332237.1 | NP_001321915.1 | 2 - 3 | 459 | 127 | |
NM_101522.4 | NP_564002.1 | 3 - 4 | 404 | 127 | ||
GenBank mRNA | AF372829.1 | - | 3 - 4 | 225 | 127 | |
AK175235.1 | - | 3 - 4 | 283 | 127 | ||
BT024601.1 | - | 3 - 4 | 225 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |