Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198086.1 | NP_001185015.1 | 15 - 16 | 4614 | 144 | |
NM_001332245.1 | NP_001321740.1 | 15 - 16 | 4854 | 144 | ||
NM_001332246.1 | NP_001321737.1 | 15 - 16 | 5064 | 144 | ||
NM_001332247.1 | NP_001321739.1 | 15 - 16 | 4947 | 144 | ||
NM_001332248.1 | NP_001321738.1 | 16 - 17 | 4791 | 144 | ||
NM_001332249.1 | NP_001321742.1 | 15 - 16 | 4552 | 144 | ||
NM_001332250.1 | NP_001321741.1 | 15 - 16 | 4620 | 144 | ||
NM_001332251.1 | NP_001321736.1 | 15 - 16 | 4900 | 144 | ||
NM_101532.3 | NP_173115.1 | 14 - 15 | 4647 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |