Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198089.1 | NP_001185018.1 | 8 - 9 | 1617 | 83 | |
NM_001332274.1 | NP_001321275.1 | 7 - 8 | 1664 | 83 | ||
NM_101564.4 | NP_173147.2 | 7 - 8 | 1664 | 83 | ||
GenBank mRNA | BT046180.1 | - | 7 - 8 | 1326 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |