Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198090.2 | NP_001185019.1 | 11 - 12 | 2335 | 95 | |
NM_001332280.1 | NP_001322494.1 | 11 - 12 | 2461 | 95 | ||
NM_101571.2 | NP_564014.1 | 11 - 12 | 2278 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |